Anti-ATP5A1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9777CEA 448 CHF
ABGEAP9777C
Anti-ATP5A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATP5A1 (ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR