Anti-SUPV3L1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9772AEA 407 CHF
ABGEAP9772A
Anti-SUPV3L1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SUPV3L1 (suppressor of var1, 3-like 1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR