Anti-NDUFS2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9769CEA 419 CHF
ABGEAP9769C
Anti-NDUFS2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFS2 (NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR