Anti-HSD11B2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9764CEA 407 CHF
ABGEAP9764C
Anti-HSD11B2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSD11B2 (hydroxysteroid (11-beta) dehydrogenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR