Anti-ZNF473 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9752AEA 419 CHF
ABGEAP9752A
Anti-ZNF473 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF473 (zinc finger protein 473)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR