Anti-PIGN Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9751AEA 378 CHF
ABGEAP9751A
Anti-PIGN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PIGN (phosphatidylinositol glycan anchor biosynthesis, class N)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR