Anti-MAPK1IP1L Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9730CEA 407 CHF
ABGEAP9730C
Anti-MAPK1IP1L Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAPK1IP1L (mitogen-activated protein kinase 1 interacting protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR