Anti-PDE6C Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9728CEA 419 CHF
ABGEAP9728C
Anti-PDE6C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDE6C (phosphodiesterase 6C, cGMP-specific, cone, alpha prime)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR