Anti-UPK1B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9725CEA 407 CHF
ABGEAP9725C
Anti-UPK1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UPK1B (uroplakin 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR