Anti-SIRT3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9723AEA 378 CHF
ABGEAP9723A
Anti-SIRT3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADPRHL2 (ADP-ribosylhydrolase like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR