Anti-STRIP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9717CEA 405 CHF
ABGEAP9717C
Anti-STRIP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STRIP1 (striatin interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR