Anti-ADH7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9703BEA 405 CHF
ABGEAP9703B
Anti-ADH7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADH7 (alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR