Anti-PDHA1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9652DEA 378 CHF
ABGEAP9652D
Anti-PDHA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDHA1 (pyruvate dehydrogenase (lipoamide) alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR