Anti-NPW Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9644BEA 378 CHF
ABGEAP9644B
Anti-NPW Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NPW (neuropeptide W)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR