Anti-COX6A2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9616CEA 431 CHF
ABGEAP9616C
Anti-COX6A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX6A2 (cytochrome c oxidase subunit VIa polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR