Anti-CBX2 Rabbit Polyclonal Antibody [clone: 429CT16.3.1]

Supplier: Abgent
ABGEAP9605AEA 407 CHF
ABGEAP9605A
Anti-CBX2 Rabbit Polyclonal Antibody [clone: 429CT16.3.1]
Antibodies
Type: Primary
Antigen: CBX2 (chromobox homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR