Anti-WDR83 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9575CEA 378 CHF
ABGEAP9575C
Anti-WDR83 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WDR83 (WD repeat domain 83)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR