Anti-KATNBL1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9571AEA 448 CHF
ABGEAP9571A
Anti-KATNBL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KATNBL1 (katanin p80 subunit B-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR