Anti-KIF6 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9564BEA 407 CHF
ABGEAP9564B
Anti-KIF6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIF6 (kinesin family member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR