Anti-LUC7L2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9549AEA 407 CHF
ABGEAP9549A
Anti-LUC7L2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LUC7L2 (LUC7-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR