Anti-NXPH1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9543BEA 405 CHF
ABGEAP9543B
Anti-NXPH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NXPH1 (neurexophilin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR