Anti-LRRC4 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9530BEA 448 CHF
ABGEAP9530B
Anti-LRRC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LRRC4 (leucine rich repeat containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR