Anti-DIO3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9529BEA 405 CHF
ABGEAP9529B
Anti-DIO3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DIO3 (deiodinase, iodothyronine, type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR