Anti-TNFRSF10D Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9526CEA 448 CHF
ABGEAP9526C
Anti-TNFRSF10D Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNFRSF10D (tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR