Anti-FANCC Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9522BEA 405 CHF
ABGEAP9522B
Anti-FANCC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FANCC (Fanconi anemia, complementation group C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR