Anti-AMY1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9519CEA 405 CHF
ABGEAP9519C
Anti-AMY1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AMY1A (amylase, alpha 1A (salivary))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR