Anti-GBE1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9506CEA 407 CHF
ABGEAP9506C
Anti-GBE1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GBE1 (glucan (1,4-alpha-), branching enzyme 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR