Anti-IRX2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9491CEA 448 CHF
ABGEAP9491C
Anti-IRX2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRX2 (iroquois homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR