Anti-BRINP3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9487AEA 407 CHF
ABGEAP9487A
Anti-BRINP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BRINP3 (bone morphogenetic protein/retinoic acid inducible neural-specific 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR