Anti-PREX1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9461CEA 378 CHF
ABGEAP9461C
Anti-PREX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PREX1 (phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR