Anti-ELF3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9441BEA 378 CHF
ABGEAP9441B
Anti-ELF3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ELF3 (E74-like factor 3 (ets domain transcription factor, epithelial-specific ))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR