Anti-HNF4A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9435AEA 378 CHF
ABGEAP9435A
Anti-HNF4A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HNF4A (hepatocyte nuclear factor 4, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR