Anti-GSTA2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9413AEA 405 CHF
ABGEAP9413A
Anti-GSTA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSTA2 (glutathione S-transferase alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR