Anti-DIAPH2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9407CEA 378 CHF
ABGEAP9407C
Anti-DIAPH2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DIAPH2 (diaphanous-related formin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR