Anti-KDM4B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9393AEA 407 CHF
ABGEAP9393A
Anti-KDM4B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KDM4B (lysine (K)-specific demethylase 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR