Anti-LIPE Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9376BEA 407 CHF
ABGEAP9376B
Anti-LIPE Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LIPE (lipase, hormone-sensitive)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR