Anti-GPR50 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9343CEA 405 CHF
ABGEAP9343C
Anti-GPR50 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPR50 (G protein-coupled receptor 50)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR