Anti-BCL3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9337CEA 378 CHF
ABGEAP9337C
Anti-BCL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BCL3 (B-cell CLL/lymphoma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR