Anti-EYA4 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9330BEA 448 CHF
ABGEAP9330B
Anti-EYA4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EYA4 (eyes absent homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR