Anti-CENPK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9316BEA 407 CHF
ABGEAP9316B
Anti-CENPK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CENPK (centromere protein K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR