Anti-FLT1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9312AEA 378 CHF
ABGEAP9312A
Anti-FLT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR