Anti-SH3BGRL3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9302CEA 448 CHF
ABGEAP9302C
Anti-SH3BGRL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SH3BGRL3 (SH3 domain binding glutamate-rich protein like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR