Anti-GABRA4 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9298CEA 448 CHF
ABGEAP9298C
Anti-GABRA4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GABRA4 (gamma-aminobutyric acid (GABA) A receptor, alpha 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR