Anti-ACSM1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9284AEA 448 CHF
ABGEAP9284A
Anti-ACSM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACSM1 (acyl-CoA synthetase medium-chain family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR