Anti-PTMS Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9279BEA 405 CHF
ABGEAP9279B
Anti-PTMS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTMS (parathymosin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR