Anti-TSPEAR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9275CEA 419 CHF
ABGEAP9275C
Anti-TSPEAR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TSPEAR (thrombospondin-type laminin G domain and EAR repeats)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR