Anti-KCTD12 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9273CEA 407 CHF
ABGEAP9273C
Anti-KCTD12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCTD12 (potassium channel tetramerization domain containing 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR