Anti-ATP1B2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9271CEA 378 CHF
ABGEAP9271C
Anti-ATP1B2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATP1B2 (ATPase, Na+/K+ transporting, beta 2 polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR