Anti-TOP2A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9248BEA 419 CHF
ABGEAP9248B
Anti-TOP2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR