Anti-MALD2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9219BEA 407 CHF
ABGEAP9219B
Anti-MALD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MALD2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR